2-Amino-1,3-propanediol, 98%
2-Amino-1,3-propanediol, 98%
2-Amino-1,3-propanediol, 98%
2-Amino-1,3-propanediol, 98%
Thermo Scientific Chemicals
2-Amino-1,3-propanediol, 98%
CAS: 534-03-2 | C3H9NO2 | 91.11 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
1 g
5 g
Packaging:
View all
Glass Bottle
Catalog number FSA401030250
Price (MYR)
1,836.00
EA
Add to cart
Quantity:
25 g
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
1,836.00
EA
Add to cart
Specifications
Chemical Name or Material2-Amino-1, 3-propanediol
CAS534-03-2
CAS Min %97.5
Health Hazard 1GHS Signal Word: Danger
CAS Max %100.0