1-(2-Aminoethyl)piperazine, 99%
1-(2-Aminoethyl)piperazine, 99%
1-(2-Aminoethyl)piperazine, 99%
1-(2-Aminoethyl)piperazine, 99%
Thermo Scientific Chemicals
1-(2-Aminoethyl)piperazine, 99%
CAS: 140-31-8 | C6H15N3 | 129.21 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
Packaging:
Glass bottle
Catalog number FSA400765000
Price (MYR)
451.00
BOT
Add to cart
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
451.00
BOT
Add to cart
Specifications
Chemical Name or Material1-(2-Aminoethyl)piperazine
Name Note99%
CAS140-31-8
CAS Min %98.5
Health Hazard 1GHS Signal Word: Danger