4-(2-Aminoethyl)morpholine, 98+%
4-(2-Aminoethyl)morpholine, 98+%
4-(2-Aminoethyl)morpholine, 98+%
4-(2-Aminoethyl)morpholine, 98+%
Thermo Scientific Chemicals
4-(2-Aminoethyl)morpholine, 98+%
CAS: 2038-03-1 | C6H14N2O | 130.19 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass bottle
Catalog number FSA400750050
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or Material4-(2-Aminoethyl)morpholine
CAS2038-03-1
CAS Min %98.5
Health Hazard 1GHS Signal Word: Danger
CAS Max %100.0