Agarose medium EEO, for electrophoresis, (Mr = 0.16 - 0.19)
Agarose medium EEO, for electrophoresis, (Mr = 0.16 - 0.19)
Agarose medium EEO, for electrophoresis, (Mr = 0.16 - 0.19)
Agarose medium EEO, for electrophoresis, (Mr = 0.16 - 0.19)
Thermo Scientific Chemicals
Agarose medium EEO, for electrophoresis, (Mr = 0.16 - 0.19)
CAS: 9012-36-6
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
Packaging:
Glass Bottle
Catalog number FSA400420250
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Packaging:
Glass Bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialAgarose medium EEO
Physical FormFine Crystalline Powder
CAS9012-36-6
Melting Point86.5°C to 89.5°C (1.5% gel)
ColorCream to White