Agarose medium-low EEO, for electrophoresis, (Mr = 0.12)
CAS: 9012-36-6
Have Questions?
Change view
Quantity:
500 g
25 g
100 g
Packaging:
Glass Bottle
Catalog number FSA400415000
Price (MYR)
3,489.00
EA
Add to cart
Quantity:
500 g
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
3,489.00
EA
Add to cart
Specifications
Chemical Name or MaterialAgarose medium-low EEO
Physical FormFine Crystalline Powder
CAS9012-36-6
Melting Point86.5°C to 89.5°C (1.5% gel)
ColorCream to White