Acetylcholine iodide, 99%
Acetylcholine iodide, 99%
Acetylcholine iodide, 99%
Acetylcholine iodide, 99%
Thermo Scientific Chemicals
Acetylcholine iodide, 99%
CAS: 2260-50-6 | C7H16INO2 | 273.11 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
10 g
Catalog number FSA400180500
Price (MYR)
3,329.00
G
Add to cart
Quantity:
50 g
Request bulk or custom format
Price (MYR)
3,329.00
G
Add to cart
Specifications
Chemical Name or MaterialAcetylcholine iodide
Name Note99%
CAS2260-50-6
CAS Min %98.5
Health Hazard 1GHS Signal Word: Warning