2-Chloro-3-quinolinecarboxaldehyde, 98%
2-Chloro-3-quinolinecarboxaldehyde, 98%
2-Chloro-3-quinolinecarboxaldehyde, 98%
2-Chloro-3-quinolinecarboxaldehyde, 98%
2-Chloro-3-quinolinecarboxaldehyde, 98%
CAS: 73568-25-9 | C10H6ClNO | 191.62 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
Packaging:
Glass bottle
Catalog number FSA399550010
Price (MYR)
-
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or Material2-chloro-3-quinolinecarboxaldehyde
CAS73568-25-9
CAS Min %97.5
Health Hazard 1GHS Signal Word: Warning
CAS Max %100.0