1-Butanol, 99+%, Extra Dry, AcroSeal™
1-Butanol, 99+%, Extra Dry, AcroSeal™
1-Butanol, 99+%, Extra Dry, AcroSeal™
1-Butanol, 99+%, Extra Dry, AcroSeal™
Thermo Scientific Chemicals
1-Butanol, 99+%, Extra Dry, AcroSeal™
CAS: 71-36-3 | C4H10O | 74.123 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
Packaging:
Glass Bottle
Catalog number FSA398960010
Price (MYR)
2,349.00
EA
Add to cart
Quantity:
1 L
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
2,349.00
EA
Add to cart
Specifications
Chemical Name or Material1-Butanol
Melting Point-89.0°C
Name NoteFor spectroscopy
CAS71-36-3
CAS Min %99.0