2-Methyl-1-propanol, 99+%, Extra Dry, AcroSeal™
2-Methyl-1-propanol, 99+%, Extra Dry, AcroSeal™
2-Methyl-1-propanol, 99+%, Extra Dry, AcroSeal™
2-Methyl-1-propanol, 99+%, Extra Dry, AcroSeal™
Thermo Scientific Chemicals
2-Methyl-1-propanol, 99+%, Extra Dry, AcroSeal™
CAS: 78-83-1 | C4H10O | 74.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
1 L
Catalog number FSA398951000
Price (MYR)
396.00
EA
Add to cart
Quantity:
100 mL
Request bulk or custom format
Price (MYR)
396.00
EA
Add to cart
Specifications
Chemical Name or Material2-Methyl-1-propanol
Name NoteExtra Dry
CAS78-83-1
CAS Min %99.0
Health Hazard 1GHS Signal Word: Danger