2-Bromo-5-fluorobenzaldehyde, 96%, Thermo Scientific Chemicals
2-Bromo-5-fluorobenzaldehyde, 96%, Thermo Scientific Chemicals
2-Bromo-5-fluorobenzaldehyde, 96%, Thermo Scientific Chemicals
2-Bromo-5-fluorobenzaldehyde, 96%, Thermo Scientific Chemicals
2-Bromo-5-fluorobenzaldehyde, 96%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Packaging:
Glass bottle
Catalog number FSA398910050
Price (MYR)
-
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or Material2-Bromo-5-fluorobenzaldehyde
CAS94569-84-3
CAS Min %95.0
Health Hazard 1GHS Signal Word: Warning
CAS Max %100.0