1-Naphthyl phosphate, disodium salt hydrate, 98%
1-Naphthyl phosphate, disodium salt hydrate, 98%
1-Naphthyl phosphate, disodium salt hydrate, 98%
1-Naphthyl phosphate, disodium salt hydrate, 98%
1-Naphthyl phosphate, disodium salt hydrate, 98%
CAS: 207569-06-0 | C10H7Na2O4P | 268.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
Packaging:
Glass bottle
Catalog number FSA398260050
Price (MYR)
-
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or Material1-Naphthyl phosphate, disodium salt hydrate
CAS207569-06-0
CAS Min %97.5
Health Hazard 1GHS Signal Word: Warning
CAS Max %100.0