Indole-4-carboxylic acid, 98%, Thermo Scientific™
Indole-4-carboxylic acid, 98%, Thermo Scientific™
Indole-4-carboxylic acid, 98%, Thermo Scientific™
Indole-4-carboxylic acid, 98%, Thermo Scientific™
Indole-4-carboxylic acid, 98%, Thermo Scientific™
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
Packaging:
Glass bottle
Catalog number FSA398060010
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialIndole-4-carboxylic acid
CAS2124-55-2
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement:
Harmful in contact with skin.
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Harmful if swallowed.
Health Hazard 3GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.