4-(Trifluoromethyl)phenylhydrazine, 96%
4-(Trifluoromethyl)phenylhydrazine, 96%
4-(Trifluoromethyl)phenylhydrazine, 96%
4-(Trifluoromethyl)phenylhydrazine, 96%
4-(Trifluoromethyl)phenylhydrazine, 96%
CAS: 368-90-1 | C7H7F3N2 | 176.14 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
Packaging:
Glass bottle
Catalog number FSA397950050
Price (MYR)
-
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or Material4-(Trifluoromethyl)phenylhydrazine
CAS368-90-1
CAS Min %95.0
Health Hazard 1GHS Signal Word: Warning
CAS Max %100.0