4-(Trifluoromethyl)benzenesulfonyl chloride, 98%
4-(Trifluoromethyl)benzenesulfonyl chloride, 98%
4-(Trifluoromethyl)benzenesulfonyl chloride, 98%
4-(Trifluoromethyl)benzenesulfonyl chloride, 98%
Thermo Scientific Chemicals
4-(Trifluoromethyl)benzenesulfonyl chloride, 98%
CAS: 2991-42-6 | C7H4ClF3O2S | 244.61 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Packaging:
Glass bottle
Catalog number FSA397760050
Price (MYR)
596.00
EA
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
596.00
EA
Add to cart
Specifications
Chemical Name or Material4-(Trifluoromethyl)benzenesulfonyl chloride
CAS2991-42-6
CAS Min %97.5
Health Hazard 1GHS Signal Word: Danger
CAS Max %100.0