(Trimethylsilyl)diazomethane, approx. 2M solution in diethyl ether
CAS: 18107-18-1 | C4H10N2Si | 114.23 g/mol
Have Questions?
Change view
Quantity:
100 mL
5 mL
Packaging:
Glass bottle
Catalog number FSA397621000
Price (MYR)
-
Quantity:
100 mL
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or Material(Trimethylsilyl)diazomethane
Name NoteApprox. 2M solution in diethyl ether
CAS18107-18-1
CAS Min %28.19
Health Hazard 1GHS Signal Word: Danger