Methyl sulfoxide, 99.8+%, for HPLC, Thermo Scientific Chemicals
Methyl sulfoxide, 99.8+%, for HPLC, Thermo Scientific Chemicals
Methyl sulfoxide, 99.8+%, for HPLC, Thermo Scientific Chemicals
Methyl sulfoxide, 99.8+%, for HPLC, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Methyl sulfoxide, 99.8+%, for HPLC, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
Catalog number FSA397600010
Price (MYR)
952.00
EA
Add to cart
Quantity:
1 L
Request bulk or custom format
Price (MYR)
952.00
EA
Add to cart
Specifications
Chemical Name or MaterialMethyl sulfoxide
Melting Point18.4°C
Name NoteFor HPLC
CAS67-68-5
Density1.1000g/mL