2,6-Dimethylbenzaldehyde, 97%
2,6-Dimethylbenzaldehyde, 97%
2,6-Dimethylbenzaldehyde, 97%
2,6-Dimethylbenzaldehyde, 97%
Thermo Scientific Chemicals
2,6-Dimethylbenzaldehyde, 97%
CAS: 1123-56-4 | C9H10O | 134.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Packaging:
Glass bottle
Catalog number FSA397560050
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or Material2, 6-Dimethylbenzaldehyde
CAS1123-56-4
CAS Min %96.0
Health Hazard 1GHS Signal Word: Warning
CAS Max %100.0