3-Bromobenzoyl chloride, 97%
3-Bromobenzoyl chloride, 97%
3-Bromobenzoyl chloride, 97%
3-Bromobenzoyl chloride, 97%
Thermo Scientific Chemicals
3-Bromobenzoyl chloride, 97%
CAS: 1711-09-7 | C7H4BrClO | 219.46 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Packaging:
Glass bottle
Catalog number FSA397450250
Price (MYR)
892.00
EA
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
892.00
EA
Add to cart
Specifications
Chemical Name or Material3-Bromobenzoyl chloride
CAS1711-09-7
CAS Min %96.0
Health Hazard 1GHS Signal Word: Danger
CAS Max %100.0