4-(Bromomethyl)pyridine hydrobromide, 97%
4-(Bromomethyl)pyridine hydrobromide, 97%
4-(Bromomethyl)pyridine hydrobromide, 97%
4-(Bromomethyl)pyridine hydrobromide, 97%
Thermo Scientific Chemicals
4-(Bromomethyl)pyridine hydrobromide, 97%
CAS: 73870-24-3 | C6H7Br2N | 252.94 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Catalog number FSA397330250
Price (MYR)
1,600.00
EA
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (MYR)
1,600.00
EA
Add to cart
Specifications
Chemical Name or Material4-(Bromomethyl)pyridine hydrobromide
CAS73870-24-3
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
Causes severe skin burns and eye damage.
Health Hazard 3GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.