Methylcyclohexane, 98+%, Extra Dry, AcroSeal™
Methylcyclohexane, 98+%, Extra Dry, AcroSeal™
Methylcyclohexane, 98+%, Extra Dry, AcroSeal™
Methylcyclohexane, 98+%, Extra Dry, AcroSeal™
Thermo Scientific Chemicals
Methylcyclohexane, 98+%, Extra Dry, AcroSeal™
CAS: 108-87-2 | C7H14 | 98.19 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
Catalog number FSA397241000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 mL
Request bulk or custom format
Specifications
Chemical Name or MaterialMethylcyclohexane
Name NoteExtra Dry
CAS108-87-2
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement:
May cause drowsiness or dizziness.
May be fatal if swallowed and enters airways.
Causes skin irritation.
Toxic to aquatic life with long lasting effects.
Highly flammable liquid and vapor.