3,4-Diaminoanisole dihydrochloride, 98%
3,4-Diaminoanisole dihydrochloride, 98%
3,4-Diaminoanisole dihydrochloride, 98%
3,4-Diaminoanisole dihydrochloride, 98%
Thermo Scientific Chemicals
3,4-Diaminoanisole dihydrochloride, 98%
CAS: 59548-39-9 | C7H10N22ClH | 211.09 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
Catalog number FSA396990050
Price (MYR)
868.00
EA
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (MYR)
868.00
EA
Add to cart
Specifications
Chemical Name or Material3, 4-Diaminoanisole dihydrochloride
CAS59548-39-9
Health Hazard 1GHS Signal Word: Warning
Melting Point206.0°C to 209.0°C
EINECS Number261-801-