Chlorobenzene, 99.8%, Extra Dry, AcroSeal™
Chlorobenzene, 99.8%, Extra Dry, AcroSeal™
Chlorobenzene, 99.8%, Extra Dry, AcroSeal™
Chlorobenzene, 99.8%, Extra Dry, AcroSeal™
Chlorobenzene, 99.8%, Extra Dry, AcroSeal™
Thermo Scientific Chemicals
Chlorobenzene, 99.8%, Extra Dry, AcroSeal™
CAS: 108-90-7 | C6H5Cl | 112.556 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
Packaging:
AcroSeal™ Glass Bottle
Catalog number FSA396971000
Price (MYR)
475.00
EA
Add to cart
Quantity:
100 mL
Packaging:
AcroSeal™ Glass Bottle
Request bulk or custom format
Price (MYR)
475.00
EA
Add to cart
Specifications
Chemical Name or MaterialChlorobenzene
Melting Point-45.0°C
Name NoteExtra Dry
CAS108-90-7