2-Methoxyethanol, 99+%, Extra Dry, AcroSeal™
2-Methoxyethanol, 99+%, Extra Dry, AcroSeal™
2-Methoxyethanol, 99+%, Extra Dry, AcroSeal™
2-Methoxyethanol, 99+%, Extra Dry, AcroSeal™
Thermo Scientific Chemicals
2-Methoxyethanol, 99+%, Extra Dry, AcroSeal™
CAS: 109-86-4 | C3H8O2 | 76.09 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
1 L
Catalog number FSA396891000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 mL
Request bulk or custom format
Specifications
Chemical Name or Material2-Methoxyethanol
Name NoteExtra Dry
CAS109-86-4
CAS Min %99.0
Health Hazard 1GHS Signal Word: Danger