Poly(vinyl alcohol), 88% hydrolyzed, average M.W. 20000-30000
Poly(vinyl alcohol), 88% hydrolyzed, average M.W. 20000-30000
Poly(vinyl alcohol), 88% hydrolyzed, average M.W. 20000-30000
Poly(vinyl alcohol), 88% hydrolyzed, average M.W. 20000-30000
Thermo Scientific Chemicals
Poly(vinyl alcohol), 88% hydrolyzed, average M.W. 20000-30000
CAS: 9002-89-5 | (C2H4O)n | 44.05 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
25 g
1 kg
Packaging:
Plastic bottle
Catalog number FSA396765000
Price (MYR)
679.00
EA
Add to cart
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
679.00
EA
Add to cart
Specifications
Chemical Name or MaterialPoly(vinyl alcohol)
ColorWhite to Yellow
pH4.5 to 6.5 (4% aq. soln.)
CAS9002-89-5
Physical FormPowder
View more
Indicator
RUO – Research Use Only