1-Nonanesulfonic acid, sodium salt, 99+%, Ion pair chromatography, anhydrous, Thermo Scientific Chemicals
1-Nonanesulfonic acid, sodium salt, 99+%, Ion pair chromatography, anhydrous, Thermo Scientific Chemicals
1-Nonanesulfonic acid, sodium salt, 99+%, Ion pair chromatography, anhydrous, Thermo Scientific Chemicals
1-Nonanesulfonic acid, sodium salt, 99+%, Ion pair chromatography, anhydrous, Thermo Scientific Chemicals
1-Nonanesulfonic acid, sodium salt, 99+%, Ion pair chromatography, anhydrous, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
Packaging:
Glass bottle
Catalog number FSA396700250
Price (MYR)
-
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or Material1-Nonanesulfonic acid, sodium salt
Name NoteAnhydrous
CAS35192-74-6
Absorbance0.02 max. (C=0.2 M in water) at 230nm, 0.02 max. (C=0.2 M in water) at 260nm, 0.03 max. (C=0.2 M in water) at 220nm, 0.05 max. (C=0.2 M in water) at 210nm
CAS Min %99.0
View more
For ion pair chromatography
RUO – Research Use Only