Activated charcoal, NORIT™ GAC 1240, 12-40 mesh
Activated charcoal, NORIT™ GAC 1240, 12-40 mesh
Activated charcoal, NORIT™ GAC 1240, 12-40 mesh
Activated charcoal, NORIT™ GAC 1240, 12-40 mesh
Thermo Scientific Chemicals
Activated charcoal, NORIT™ GAC 1240, 12-40 mesh
CAS: 7440-44-0 | C | 12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
Packaging:
Glass bottle
Catalog number FSA395712500
Price (MYR)
440.00
BOT
Add to cart
Quantity:
250 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
440.00
BOT
Add to cart
Specifications
Chemical Name or MaterialActivated charcoal,NORIT* GAC 124
CAS7440-44-0
Melting Point3652.0°C
ColorBlack
EINECS Number231-153-3
View more
Filter aid
RUO – Research Use Only