2-chlorophenylacetyl Chloride, 97%
2-chlorophenylacetyl Chloride, 97%
2-chlorophenylacetyl Chloride, 97%
2-chlorophenylacetyl Chloride, 97%
Thermo Scientific Chemicals
2-chlorophenylacetyl Chloride, 97%
CAS: 51512-09-5 | C8H6Cl2O | 189.04 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
Packaging:
Glass bottle
Catalog number FSA395080250
Price (MYR)
609.00
BOT
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
609.00
BOT
Add to cart
Specifications
Chemical Name or Material2-chlorophenylacetyl chloride
CAS51512-09-5
CAS Min %96.0
ColorYellow
Health Hazard 1GHS Signal Word: Danger