N,N-Diethyl-p-phenylenediamine sulfate, 99%
N,N-Diethyl-p-phenylenediamine sulfate, 99%
N,N-Diethyl-p-phenylenediamine sulfate, 99%
N,N-Diethyl-p-phenylenediamine sulfate, 99%
Thermo Scientific Chemicals
N,N-Diethyl-p-phenylenediamine sulfate, 99%
N, N-Diethyl-p-phenylenediamine sulfate, 99%, C10H16N2.H2SO4, CAS Number-6283-63-2 | CAS: 6283-63-2 | C10H16N2·H2SO4 | 262.33 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
50 g
1 kg
Packaging:
Glass bottle
Catalog number FSA395002500
Price (MYR)
663.00
BOT
Add to cart
Quantity:
250 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
663.00
BOT
Add to cart
Specifications
Chemical Name or MaterialN, N-Diethyl-p-phenylenediamine sulfate
CAS6283-63-2
CAS Min %98.5
Health Hazard 1GHS Signal Word: Warning
CAS Max %100.0