4-Methoxypiperidine, 98+%
4-Methoxypiperidine, 98+%
4-Methoxypiperidine, 98+%
4-Methoxypiperidine, 98+%
Thermo Scientific Chemicals
4-Methoxypiperidine, 98+%
CAS: 4045-24-3 | C6H13NO | 115.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
Packaging:
Glass bottle
Catalog number FSA393390050
Price (MYR)
2,469.00
EA
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
2,469.00
EA
Add to cart
Specifications
Chemical Name or Material4-Methoxypiperidine
CAS4045-24-3
CAS Min %98.0
Health Hazard 1GHS Signal Word: Danger
CAS Max %100.0