Potassium hydroxide, 1N solution in ethanol
Potassium hydroxide, 1N solution in ethanol
Potassium hydroxide, 1N solution in ethanol
Potassium hydroxide, 1N solution in ethanol
Potassium hydroxide, 1N solution in ethanol
Thermo Scientific Chemicals
Potassium hydroxide, 1N solution in ethanol
CAS: 1310-58-3 | HKO | 56.11 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
1 L
Packaging:
Plastic bottle
Catalog number FSA393322500
Price (MYR)
244.00
EA
Add to cart
Quantity:
250 mL
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
244.00
EA
Add to cart
Chemical Identifiers
CAS1310-58-3, 64-17-5
Specifications
Chemical Name or MaterialPotassium hydroxide
ColorBrown-Orange to Yellow
Name Note1N solution in ethanol
Boiling Point78.0°C
CAS1310-58-3
View more
Suitable for battery materials development.
For Manufacturing and Laboratory Use. Not for beverage use