Sand, for analysis, 40-100 mesh
Sand, for analysis, 40-100 mesh
Sand, for analysis, 40-100 mesh
Sand, for analysis, 40-100 mesh
Thermo Scientific Chemicals
Sand, for analysis, 40-100 mesh
CAS: 14808-60-7 | O2Si | 60.08 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
5 kg
Packaging:
Plastic bottle
Catalog number FSA393302500
Price (MYR)
330.00
BOT
Add to cart
Quantity:
250 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
330.00
BOT
Add to cart
Specifications
Boiling Point2230.0°C
Chemical Name or MaterialSand
Melting Point1610.0°C
CAS14808-60-7
ColorBeige to Gray
View more
Filter aid
RUO – Research Use Only