2-(2-Bromophenyl)-1,3-dioxolane, 98%
2-(2-Bromophenyl)-1,3-dioxolane, 98%
2-(2-Bromophenyl)-1,3-dioxolane, 98%
2-(2-Bromophenyl)-1,3-dioxolane, 98%
Thermo Scientific Chemicals
2-(2-Bromophenyl)-1,3-dioxolane, 98%
CAS: 34824-58-3 | C9H9BrO2 | 229.07 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
Packaging:
Glass bottle
Catalog number FSA393240250
Price (MYR)
1,599.00
EA
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,599.00
EA
Add to cart
Specifications
Chemical Name or Material2-(2-Bromophenyl)-1, 3-dioxolane
CAS34824-58-3
CAS Min %97.5
Health Hazard 1GHS Signal Word: Warning
CAS Max %100.0