Ammonium chloride, for analysis
Ammonium chloride, for analysis
Ammonium chloride, for analysis
Ammonium chloride, for analysis
Thermo Scientific Chemicals
Ammonium chloride, for analysis
CAS: 12125-02-9 | ClH4N | 53.49 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
Packaging:
View all
Glass bottle
Catalog number FSA393182500
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialAmmonium chloride
Melting Point340.0°C
CAS12125-02-9
ColorWhite
Health Hazard 1GHS Signal Word: Warning