3,5-Dibromobenzaldehyde, 98+%
3,5-Dibromobenzaldehyde, 98+%
3,5-Dibromobenzaldehyde, 98+%
3,5-Dibromobenzaldehyde, 98+%
Thermo Scientific Chemicals
3,5-Dibromobenzaldehyde, 98+%
CAS: 56990-02-4 | C7H4Br2O | 263.91 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
Packaging:
Glass bottle
Catalog number FSA393150010
Price (MYR)
332.00
EA
Add to cart
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
332.00
EA
Add to cart
Specifications
Chemical Name or Material3, 5-Dibromobenzaldehyde
CAS56990-02-4
CAS Min %98.0
Health Hazard 1GHS Signal Word: Danger
CAS Max %100.0