2-Amino-5-methylthiazole, 98%
2-Amino-5-methylthiazole, 98%
2-Amino-5-methylthiazole, 98%
2-Amino-5-methylthiazole, 98%
Thermo Scientific Chemicals
2-Amino-5-methylthiazole, 98%
CAS: 7305-71-7 | C4H6N2S | 114.17 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
5 g
25 g
Packaging:
Glass bottle
Catalog number FSA392791000
Price (MYR)
1,404.00
BOT
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,404.00
BOT
Add to cart
Specifications
Chemical Name or Material2-Amino-5-methylthiazole
CAS7305-71-7
CAS Min %97.5
Health Hazard 1GHS Signal Word: Warning
CAS Max %100.0