2-Hydroxy-5-methylbenzaldehyde, 99%
2-Hydroxy-5-methylbenzaldehyde, 99%
2-Hydroxy-5-methylbenzaldehyde, 99%
2-Hydroxy-5-methylbenzaldehyde, 99%
Thermo Scientific Chemicals
2-Hydroxy-5-methylbenzaldehyde, 99%
CAS: 613-84-3 | C8H8O2 | 136.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Packaging:
Glass bottle
Catalog number FSA392630050
Price (MYR)
1,076.00
EA
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,076.00
EA
Add to cart
Specifications
Chemical Name or Material2-Hydroxy-5-methylbenzaldehyde
CAS613-84-3
CAS Min %98.5
Health Hazard 1GHS Signal Word: Warning
CAS Max %100.0