3,7-Dimethyl-1-octanol, 95%
3,7-Dimethyl-1-octanol, 95%
3,7-Dimethyl-1-octanol, 95%
3,7-Dimethyl-1-octanol, 95%
Thermo Scientific Chemicals
3,7-Dimethyl-1-octanol, 95%
CAS: 106-21-8 | C10H22O | 158.28 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 mL
Packaging:
Glass bottle
Catalog number FSA392620500
Price (MYR)
341.00
EA
Add to cart
Quantity:
50 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
341.00
EA
Add to cart
Specifications
Chemical Name or Material3, 7-Dimethyl-1-octanol
CAS106-21-8
CAS Min %94.0
Health Hazard 1GHS Signal Word: Warning
CAS Max %100.0