4-Iodobiphenyl, 97+%
4-Iodobiphenyl, 97+%
4-Iodobiphenyl, 97+%
4-Iodobiphenyl, 97+%
Thermo Scientific Chemicals
4-Iodobiphenyl, 97+%
CAS: 1591-31-7 | C12H9I | 280.11 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Packaging:
Glass bottle
Catalog number FSA392390050
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or Material4-Iodobiphenyl
CAS1591-31-7
CAS Min %97.0
Health Hazard 1GHS Signal Word: Warning
CAS Max %100.0