Silica gel orange, for drying purposes, non toxic grade, 2-5 mm
Silica gel orange, for drying purposes, non toxic grade, 2-5 mm
Silica gel orange, for drying purposes, non toxic grade, 2-5 mm
Silica gel orange, for drying purposes, non toxic grade, 2-5 mm
Thermo Scientific Chemicals
Silica gel orange, for drying purposes, non toxic grade, 2-5 mm
CAS: 1327-36-2 | Al2O5Si | 162.04 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
Packaging:
Glass Bottle
Catalog number FSA392030010
Price (MYR)
495.00
EA
Add to cart
Quantity:
1 kg
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
495.00
EA
Add to cart
Specifications
Chemical Name or MaterialSilica gel orange
ColorOrange
Physical FormBeads
CAS1327-36-2
MaterialSilica Gel
View more
Drying agent
RUO – Research Use Only