Citric acid, trisodium salt, 98%, pure, anhydrous
Citric acid, trisodium salt, 98%, pure, anhydrous
Citric acid, trisodium salt, 98%, pure, anhydrous
Citric acid, trisodium salt, 98%, pure, anhydrous
Thermo Scientific Chemicals
Citric acid, trisodium salt, 98%, pure, anhydrous
CAS: 68-04-2 | C6H5Na3O7 | 258.07 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
2.5 kg
Packaging:
Plastic bottle
Catalog number FSA391975000
Price (MYR)
336.00
BOT
Add to cart
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
336.00
BOT
Add to cart
Specifications
Chemical Name or MaterialCitric acid, trisodium salt
CAS68-04-2
Melting Point300.0°C
ColorWhite
Flash Point290°C