Benzyl alcohol, for analysis
Benzyl alcohol, for analysis
Benzyl alcohol, for analysis
Benzyl alcohol, for analysis
Thermo Scientific Chemicals
Benzyl alcohol, for analysis
CAS: 100-51-6 | C7H8O | 108.14 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
1 L
Packaging:
Glass bottle
Catalog number FSA391220025
Price (MYR)
658.00
BOT
Add to cart
Quantity:
2.5 L
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
658.00
BOT
Add to cart
Specifications
Chemical Name or MaterialBenzyl alcohol
Alkalinity0.002 meq/g max.
Acidity0.001 meq/g max.
CAS100-51-6
Health Hazard 1GHS Signal Word: Warning
View more
For analysis
RUO – Research Use Only