4-Diphenylaminobenzaldehyde, 98%, Thermo Scientific Chemicals
4-Diphenylaminobenzaldehyde, 98%, Thermo Scientific Chemicals
4-Diphenylaminobenzaldehyde, 98%, Thermo Scientific Chemicals
4-Diphenylaminobenzaldehyde, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
4-Diphenylaminobenzaldehyde, 98%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
Packaging:
Glass bottle
Catalog number FSA391180050
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or Material4-Diphenylaminobenzaldehyde
CAS4181-05-9
CAS Min %97.5
Health Hazard 1GHS Signal Word: Warning
CAS Max %100.0