3-Pyridinepropionic acid, 97%
3-Pyridinepropionic acid, 97%
3-Pyridinepropionic acid, 97%
3-Pyridinepropionic acid, 97%
Thermo Scientific Chemicals
3-Pyridinepropionic acid, 97%
CAS: 3724-19-4 | C8H9NO2 | 151.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
Packaging:
Glass bottle
Catalog number FSA390980050
Price (MYR)
1,194.00
BOT
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,194.00
BOT
Add to cart
Specifications
Chemical Name or Material3-Pyridinepropionic acid
CAS3724-19-4
CAS Min %96.0
Health Hazard 1GHS Signal Word: Warning
CAS Max %100.0