Chloroform, 99+%, for HPLC, stabilized with ethanol
Chloroform, 99+%, for HPLC, stabilized with ethanol
Chloroform, 99+%, for HPLC, stabilized with ethanol
Chloroform, 99+%, for HPLC, stabilized with ethanol
Thermo Scientific Chemicals
Chloroform, 99+%, for HPLC, stabilized with ethanol
Chloroform, >-99%, CHCl3, CAS Number-67-66-3, 64-17-5, trichloromethane, trichloroform, trichlormethan, methyl trichloride, cloroformio, methenyl trichloride, formyl trichloride, methane, trichloro, chloroforme, methane trichloride, 2.5L | CAS: 67-66-3 | CHCl3 | 119.37 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
1 L
Packaging:
Glass Bottle
Catalog number FSA390760025
Price (MYR)
846.00
BOT
Add to cart
Quantity:
2.5 L
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
846.00
BOT
Add to cart
Specifications
Chemical Name or MaterialChloroform
Melting Point-63.0°C
Name NoteStabilized with ethanol
Absorbance0.009 max. at 300nm, 0.046 max. at 265nm, 0.301 max. at 250nm, 0.699 max. at 245nm, 1 cm cell vs water, 1 cm cell vs water at 0nm
CAS67-66-3
View more
For HPLC
RUO – Research Use Only