Methyl sulfoxide, 99.5+%, for analysis, Thermo Scientific Chemicals
Methyl sulfoxide, 99.5+%, for analysis, Thermo Scientific Chemicals
Methyl sulfoxide, 99.5+%, for analysis, Thermo Scientific Chemicals
Methyl sulfoxide, 99.5+%, for analysis, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Methyl sulfoxide, 99.5+%, for analysis, Thermo Scientific Chemicals
Have Questions?
Catalog NumberQuantityPackaging
FSA3907500252.5 LGlass Bottle
Catalog number FSA390750025
Price (MYR)
1,031.00
EA
Add to cart
Quantity:
2.5 L
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
1,031.00
EA
Add to cart
Specifications
Chemical Name or MaterialMethyl sulfoxide
Melting Point18.4°C
CAS67-68-5
Acidity0.0002 meq/g max.
Density1.1000g/mL
View more
For analysis
RUO – Research Use Only