Dichloromethane, 99.5%, for analysis, stabilized with ethanol
Dichloromethane, 99.5%, for analysis, stabilized with ethanol
Dichloromethane, 99.5%, for analysis, stabilized with ethanol
Dichloromethane, 99.5%, for analysis, stabilized with ethanol
Thermo Scientific Chemicals
Dichloromethane, 99.5%, for analysis, stabilized with ethanol
CAS: 75-09-2 | CH2Cl2 | 84.93 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
Packaging:
Glass Bottle
Catalog number FSA390700010
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 L
Packaging:
Glass Bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialDichloromethane
Melting Point-97.0°C
CAS75-09-2
Health Hazard 1GHS Signal Word: Warning
Acidity0.0005 meq/g max.