Isopropyl ether, for analysis, stabilized
Isopropyl ether, for analysis, stabilized
Isopropyl ether, for analysis, stabilized
Isopropyl ether, for analysis, stabilized
Thermo Scientific Chemicals
Isopropyl ether, for analysis, stabilized
CAS: 108-20-3 | C6H14O | 102.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
2.5 L
Packaging:
Glass bottle
Catalog number FSA390660010
Price (MYR)
436.00
BOT
Add to cart
Quantity:
1 L
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
436.00
BOT
Add to cart
Specifications
Chemical Name or MaterialIsopropyl ether
Acidity0.0004 meq/g max.
CAS108-20-3
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement:
May cause drowsiness or dizziness.
Highly flammable liquid and vapor.
May form explosive peroxides.
Repeated exposure may cause skin dryness or cracking.
View more
For analysis
RUO – Research Use Only