tert-Butyl methyl ether, for analysis
tert-Butyl methyl ether, for analysis
tert-Butyl methyl ether, for analysis
tert-Butyl methyl ether, for analysis
Thermo Scientific Chemicals
tert-Butyl methyl ether, for analysis
CAS: 1634-04-4 | C5H12O | 88.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
Packaging:
Glass Bottle
Catalog number FSA390480010
Price (MYR)
359.00
EA
Add to cart
Quantity:
1 L
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
359.00
EA
Add to cart
Specifications
Chemical Name or Materialtert-Butyl methyl ether
Melting Point-110.0°C
CAS1634-04-4
ColorColorless
Health Hazard 1GHS Signal Word: Danger