Calcium citrate tribasic tetrahydrate, 98+%, for analysis
Calcium citrate tribasic tetrahydrate, 98+%, for analysis
Calcium citrate tribasic tetrahydrate, 98+%, for analysis
Calcium citrate tribasic tetrahydrate, 98+%, for analysis
Thermo Scientific Chemicals
Calcium citrate tribasic tetrahydrate, 98+%, for analysis
CAS: 5785-44-4 | C12H10Ca3O14·4H2O | 570.5 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
Packaging:
Plastic bottle
Catalog number FSA390082500
Price (MYR)
240.00
BOT
Add to cart
Quantity:
250 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
240.00
BOT
Add to cart
Specifications
Chemical Name or MaterialCalcium citrate tribasic tetrahydrate
CAS5785-44-4
Loss on Drying10.0 % to 13.3 % (150°C, 4 hrs)
Infrared SpectrumAuthentic
Assay Percent Range98.0 to 102% (on dried basis) (Complexometry)