Ammonium hydroxide, for analysis, 25% solution in water
Ammonium hydroxide, for analysis, 25% solution in water
Ammonium hydroxide, for analysis, 25% solution in water
Ammonium hydroxide, for analysis, 25% solution in water
Thermo Scientific Chemicals
Ammonium hydroxide, for analysis, 25% solution in water
CAS: 1336-21-6 | H5NO | 35.05 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
1 L
Packaging:
Glass bottle
Catalog number FSA390030025
Price (MYR)
578.00
BOT
Add to cart
Quantity:
2.5 L
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
578.00
BOT
Add to cart
Specifications
Chemical Name or MaterialAmmonium hydroxide
ColorColorless
Boiling Point38.0°C
CAS1336-21-6
Health Hazard 1Danger